Assay Details
| Gene Symbol: LOC4331489 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773528.2 | XP_015629014.1 | 4 - 5 | 573 | 78 | |
| GenBank mRNA | AB037900.1 | - | 4 - 5 | 459 | 78 | |
| AK067991.1 | - | 4 - 5 | 548 | 78 | ||
More Information
Set Membership: |
|

LOC4331489
Genomic Map