Assay Details
| Gene Symbol: LOC4331489 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773528.2 | XP_015629014.1 | 9 - 10 | 1021 | 120 | |
| GenBank mRNA | AB037900.1 | - | 9 - 10 | 907 | 120 | |
| AK067991.1 | - | 9 - 10 | 996 | 120 | ||
More Information
Set Membership: |
|

LOC4331489
Genomic Map