Assay Details
| Gene Symbol: LOC4331590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776545.2 | XP_015632031.1 | 1 - 2 | 188 | 100 | |
| GenBank mRNA | AK073999.1 | - | 1 - 2 | 142 | 100 | |
| FJ940198.1 | - | 1 - 2 | 73 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015776545.2 transcripts.
Set Membership: |
|

LOC4331590
Genomic Map