Assay Details
| Gene Symbol: LOC4331590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776545.2 | XP_015632031.1 | 4 - 5 | 508 | 82 | |
| GenBank mRNA | AK073999.1 | - | 4 - 5 | 462 | 82 | |
| FJ940198.1 | - | 4 - 5 | 393 | 82 | ||
More Information
Set Membership: |
|

LOC4331590
Genomic Map