Assay Details
| Gene Symbol: LOC4331646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772981.2 | XP_015628467.1 | 1 - 2 | 213 | 82 | |
| GenBank mRNA | AB032761.1 | - | 1 - 2 | 138 | 82 | |
| AK105389.1 | - | 1 - 2 | 187 | 82 | ||
| AK121527.1 | - | 1 - 2 | 205 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015772981.2 transcripts.
Set Membership: |
|

LOC4331646
Genomic Map