Assay Details
| Gene Symbol: LOC4331966 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775932.2 | XP_015631418.1 | 4 - 5 | 575 | 66 | |
| GenBank mRNA | AK069251.1 | - | 4 - 5 | 565 | 66 | |
| AK104243.1 | - | 4 - 5 | 564 | 66 | ||
| AK104699.1 | - | 4 - 5 | 474 | 66 | ||
| BT131485.1 | - | 4 - 5 | 553 | 66 | ||
More Information
Set Membership: |
|

LOC4331966
Genomic Map