Assay Details
| Gene Symbol: LOC4332020 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773742.2 | XP_015629228.1 | 17 - 18 | 1764 | 100 | |
| GenBank mRNA | AK100680.1 | - | 17 - 18 | 1763 | 100 | |
More Information
Set Membership: |
|

LOC4332020
Genomic Map