Assay Details
| Gene Symbol: LOC4332020 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773738.2 | XP_015629224.1 | 22 - 23 | 2511 | 72 | |
| XM_015773739.2 | XP_015629225.1 | 22 - 23 | 2488 | 72 | ||
| XM_015773740.2 | XP_015629226.1 | 22 - 23 | 2481 | 72 | ||
| XM_015773741.2 | XP_015629227.1 | 22 - 23 | 2454 | 72 | ||
| XM_015773742.2 | XP_015629228.1 | 23 - 24 | 2418 | 72 | ||
| GenBank mRNA | AK100680.1 | - | 23 - 24 | 2417 | 72 | |
More Information
Set Membership: |
|

LOC4332020
Genomic Map