Assay Details
| Gene Symbol: LOC4332020 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773738.2 | XP_015629224.1 | 24 - 25 | 2674 | 68 | |
| XM_015773739.2 | XP_015629225.1 | 24 - 25 | 2651 | 68 | ||
| XM_015773740.2 | XP_015629226.1 | 24 - 25 | 2644 | 68 | ||
| XM_015773741.2 | XP_015629227.1 | 24 - 25 | 2617 | 68 | ||
| XM_015773742.2 | XP_015629228.1 | 25 - 26 | 2581 | 68 | ||
| GenBank mRNA | AK100680.1 | - | 25 - 26 | 2580 | 68 | |
More Information
Set Membership: |
|

LOC4332020
Genomic Map