Assay Details
| Gene Symbol: LOC4332020 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773738.2 | XP_015629224.1 | 2 - 3 | 264 | 124 | |
| XM_015773739.2 | XP_015629225.1 | 2 - 3 | 241 | 124 | ||
| XM_015773740.2 | XP_015629226.1 | 2 - 3 | 264 | 124 | ||
| XM_015773741.2 | XP_015629227.1 | 2 - 3 | 264 | 124 | ||
| XM_015773742.2 | XP_015629228.1 | 2 - 3 | 267 | 124 | ||
| GenBank mRNA | AK100680.1 | - | 2 - 3 | 267 | 123 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015773738.2, XM_015773739.2, XM_015773740.2, XM_015773741.2, XM_015773742.2 transcripts.
Set Membership: |
|

LOC4332020
Genomic Map