Assay Details
| Gene Symbol: LOC4332049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774812.2 | XP_015630298.1 | 4 - 5 | 574 | 90 | |
| GenBank mRNA | AK103114.1 | - | 4 - 5 | 551 | 90 | |
More Information
Set Membership: |
|

LOC4332049
Genomic Map