Assay Details
| Gene Symbol: LOC4332049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774812.2 | XP_015630298.1 | 5 - 6 | 782 | 81 | |
| GenBank mRNA | AK103114.1 | - | 5 - 6 | 759 | 81 | |
| AY224505.1 | - | 1 - 2 | 170 | 81 | ||
More Information
Set Membership: |
|

LOC4332049
Genomic Map