Assay Details
| Gene Symbol: LOC4332049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774812.2 | XP_015630298.1 | 7 - 8 | 949 | 98 | |
| GenBank mRNA | AK103114.1 | - | 7 - 8 | 926 | 98 | |
| AY224505.1 | - | 3 - 4 | 337 | 98 | ||
More Information
Set Membership: |
|

LOC4332049
Genomic Map