Assay Details
| Gene Symbol: LOC4332341 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775825.2 | XP_015631311.1 | 3 - 4 | 1001 | 84 | |
| GenBank mRNA | AB022603.1 | - | 3 - 4 | 765 | 84 | |
| AK073501.1 | - | 2 - 3 | 297 | 84 | ||
| AK105648.1 | - | 3 - 4 | 768 | 84 | ||
More Information
Set Membership: |
|

LOC4332341
Genomic Map