Assay Details
| Gene Symbol: LOC4332341 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775825.2 | XP_015631311.1 | 4 - 5 | 1124 | 81 | |
| GenBank mRNA | AB022603.1 | - | 4 - 5 | 888 | 81 | |
| AK073501.1 | - | 3 - 4 | 420 | 81 | ||
| AK105648.1 | - | 4 - 5 | 891 | 81 | ||
More Information
Set Membership: |
|

LOC4332341
Genomic Map