Assay Details
| Gene Symbol: LOC4332341 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775825.2 | XP_015631311.1 | 8 - 9 | 1662 | 76 | |
| GenBank mRNA | AB022603.1 | - | 8 - 9 | 1426 | 76 | |
| AK073501.1 | - | 7 - 8 | 958 | 76 | ||
| AK105648.1 | - | 8 - 9 | 1429 | 76 | ||
More Information
Set Membership: |
|

LOC4332341
Genomic Map