Assay Details
| Gene Symbol: LOC4332368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776928.2 | XP_015632414.1 | 1 - 2 | 252 | 70 | |
| XM_015776929.2 | XP_015632415.1 | 1 - 2 | 253 | 70 | ||
| GenBank mRNA | AK069586.1 | - | 1 - 2 | 209 | 70 | |
More Information
Set Membership: |
|

LOC4332368
Genomic Map