Assay Details
| Gene Symbol: LOC4332368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776926.2 | XP_015632412.1 | 4 - 5 | 667 | 75 | |
| XM_015776928.2 | XP_015632414.1 | 3 - 4 | 474 | 75 | ||
| XM_015776929.2 | XP_015632415.1 | 3 - 4 | 472 | 75 | ||
| GenBank mRNA | AK069586.1 | - | 3 - 4 | 431 | 75 | |
More Information
Set Membership: |
|

LOC4332368
Genomic Map