Assay Details
| Gene Symbol: LOC4332441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775068.2 | XP_015630554.1 | 6 - 7 | 1183 | 106 | |
| GenBank mRNA | AB167397.1 | - | 7 - 8 | 1049 | 106 | |
| AK071034.1 | - | 6 - 7 | 1144 | 106 | ||
More Information
Set Membership: |
|

LOC4332441
Genomic Map