Assay Details
| Gene Symbol: LOC4332642 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777264.1 | XP_015632750.1 | 7 - 8 | 1362 | 120 | |
| GenBank mRNA | AK067474.1 | - | 7 - 8 | 1369 | 120 | |
More Information
Set Membership: |
|

LOC4332642
Genomic Map