Assay Details
| Gene Symbol: LOC4332803 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772491.2 | XP_015627977.1 | 2 - 3 | 847 | 67 | |
| XM_015772492.2 | XP_015627978.1 | 2 - 3 | 853 | 67 | ||
| GenBank mRNA | AK060603.1 | - | 2 - 3 | 342 | 67 | |
| AK102274.1 | - | 2 - 3 | 321 | 67 | ||
| AK121906.1 | - | 329 | 67 | |||
More Information
Set Membership: |
|

LOC4332803
Genomic Map