Assay Details
| Gene Symbol: LOC4332803 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772491.2 | XP_015627977.1 | 5 - 6 | 1369 | 69 | |
| XM_015772492.2 | XP_015627978.1 | 5 - 6 | 1375 | 69 | ||
| GenBank mRNA | AK060603.1 | - | 5 - 6 | 864 | 69 | |
| AK102274.1 | - | 4 - 5 | 964 | 69 | ||
More Information
Set Membership: |
|

LOC4332803
Genomic Map