Assay Details
| Gene Symbol: LOC4333033 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772756.2 | XP_015628242.1 | 2 - 3 | 850 | 108 | |
| XM_015772757.2 | XP_015628243.1 | 2 - 3 | 833 | 108 | ||
| GenBank mRNA | AK061587.1 | - | 1 - 2 | 152 | 108 | |
| AK073283.1 | - | 2 - 3 | 840 | 108 | ||
| KF274032.1 | - | 2 - 3 | 797 | 108 | ||
More Information
Set Membership: |
|

LOC4333033
Genomic Map