Assay Details
| Gene Symbol: LOC4333051 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776243.2 | XP_015631729.1 | 5 - 6 | 2354 | 118 | |
| XM_015776244.2 | XP_015631730.1 | 5 - 6 | 2338 | 118 | ||
| XM_015776245.2 | XP_015631731.1 | 5 - 6 | 2341 | 118 | ||
More Information
Set Membership: |
|

LOC4333051
Genomic Map