Assay Details
| Gene Symbol: LOC4333051 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776243.2 | XP_015631729.1 | 10 - 11 | 5396 | 78 | |
| XM_015776244.2 | XP_015631730.1 | 10 - 11 | 5380 | 78 | ||
| XM_015776245.2 | XP_015631731.1 | 10 - 11 | 5383 | 78 | ||
More Information
Set Membership: |
|

LOC4333051
Genomic Map