Assay Details
| Gene Symbol: LOC9267822 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772622.2 | XP_015628108.1 | 2 - 3 | 630 | 62 | |
| GenBank mRNA | AK066487.1 | - | 2 - 3 | 298 | 62 | |
More Information
Set Membership: |
|

LOC9267822
Genomic Map