Assay Details
| Gene Symbol: LOC9267822 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772622.2 | XP_015628108.1 | 4 - 5 | 874 | 78 | |
| GenBank mRNA | AK066487.1 | - | 4 - 5 | 542 | 78 | |
More Information
Set Membership: |
|

LOC9267822
Genomic Map