Assay Details
| Gene Symbol: LOC112936009 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK072553.1 | - | 5 - 6 | 706 | 90 | |
| AK100280.1 | - | 3 - 4 | 1253 | 90 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC112936009| Gene Symbol: LOC112936009 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK072553.1 | - | 5 - 6 | 706 | 90 | |
| AK100280.1 | - | 3 - 4 | 1253 | 90 | ||
Set Membership: |
|