Assay Details
| Gene Symbol: LOC112936009 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK072553.1 | - | 6 - 7 | 771 | 111 | |
| AK100280.1 | - | 4 - 5 | 1318 | 111 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC112936009| Gene Symbol: LOC112936009 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK072553.1 | - | 6 - 7 | 771 | 111 | |
| AK100280.1 | - | 4 - 5 | 1318 | 111 | ||
Set Membership: |
|