Assay Details
| Gene Symbol: LOC4334068 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775957.2 | XP_015631443.1 | 3 - 4 | 2503 | 80 | |
| GenBank mRNA | AK102769.1 | - | 2 - 3 | 2622 | 80 | |
More Information
Set Membership: |
|

LOC4334068
Genomic Map