Assay Details
| Gene Symbol: LOC4334176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775480.2 | XP_015630966.1 | 4 - 5 | 597 | 123 | |
| GenBank mRNA | AK106620.1 | - | 3 - 4 | 952 | 123 | |
More Information
Set Membership: |
|

LOC4334176
Genomic Map