Assay Details
| Gene Symbol: LOC4334196 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773557.2 | XP_015629043.1 | 4 - 5 | 753 | 71 | |
| GenBank mRNA | AK109047.1 | - | 4 - 5 | 628 | 71 | |
More Information
Set Membership: |
|

LOC4334196
Genomic Map