Assay Details
| Gene Symbol: LOC4334285 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775725.2 | XP_015631211.1 | 3 - 4 | 1228 | 81 | |
| GenBank mRNA | AK100507.1 | - | 4 - 5 | 1078 | 81 | |
More Information
Set Membership: |
|

LOC4334285
Genomic Map