Assay Details
| Gene Symbol: LOC4334285 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775725.2 | XP_015631211.1 | 6 - 7 | 1662 | 124 | |
| GenBank mRNA | AK100507.1 | - | 7 - 8 | 1511 | 124 | |
More Information
Set Membership: |
|

LOC4334285
Genomic Map