Assay Details
| Gene Symbol: LOC4334389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775114.2 | XP_015630600.1 | 10 - 11 | 1122 | 119 | |
| GenBank mRNA | AB109201.1 | - | 10 - 11 | 1100 | 119 | |
| AK059559.1 | - | 10 - 11 | 1033 | 119 | ||
| AK103751.1 | - | 10 - 11 | 1100 | 119 | ||
More Information
Set Membership: |
|

LOC4334389
Genomic Map