Assay Details
| Gene Symbol: LOC9268667 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777730.2 | XP_015633216.1 | 12 - 13 | 1900 | 80 | |
| GenBank mRNA | AK063420.1 | - | 9 - 10 | 1346 | 80 | |
More Information
Set Membership: |
|

LOC9268667
Genomic Map