Assay Details
Gene Symbol: LOC4334467 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775639.2 | XP_015631125.1 | 1 | 1809 | 150 | |
GenBank mRNA | AK070731.1 | - | 1 | 1555 | 150 | |
AK099212.1 | - | 1 | 1555 | 150 | ||
AK104696.1 | - | 1 | 1557 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
