Assay Details
| Gene Symbol: LOC4334606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776451.2 | XP_015631937.1 | 1 - 2 | 712 | 81 | |
| GenBank mRNA | AK101784.1 | - | 1 - 2 | 713 | 81 | |
| AK119215.1 | - | 1 - 2 | 536 | 81 | ||
More Information
Set Membership: |
|

LOC4334606
Genomic Map