Assay Details
| Gene Symbol: LOC4334649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774922.1 | XP_015630408.1 | 2 - 3 | 603 | 68 | |
| XM_015774923.1 | XP_015630409.1 | 2 - 3 | 527 | 68 | ||
| GenBank mRNA | AK065849.1 | - | 3 - 4 | 550 | 68 | |
More Information
Set Membership: |
|

LOC4334649
Genomic Map