Assay Details
| Gene Symbol: LOC4334721 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776659.2 | XP_015632145.1 | 4 - 5 | 493 | 64 | |
| XM_015776660.1 | XP_015632146.1 | 2 - 3 | 255 | 64 | ||
| GenBank mRNA | AK109847.1 | - | 4 - 5 | 353 | 64 | |
More Information
Set Membership: |
|

LOC4334721
Genomic Map