Assay Details
| Gene Symbol: LOC4334721 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776659.2 | XP_015632145.1 | 5 - 6 | 549 | 78 | |
| XM_015776660.1 | XP_015632146.1 | 3 - 4 | 311 | 78 | ||
| GenBank mRNA | AK109847.1 | - | 5 - 6 | 409 | 78 | |
More Information
Set Membership: |
|

LOC4334721
Genomic Map