Assay Details
| Gene Symbol: LOC4335113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780498.2 | XP_015635984.1 | 2 | 1671 | 89 | |
| GenBank mRNA | AY569610.1 | - | 1 | 852 | 89 | |
More Information
Set Membership: |
|

LOC4335113
Genomic Map