Assay Details
Gene Symbol: LOC4335765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780381.1 | XP_015635867.1 | 13 - 14 | 1339 | 78 | |
GenBank mRNA | AK066667.1 | - | 13 - 14 | 1337 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
