Assay Details
| Gene Symbol: LOC4335881 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781648.2 | XP_015637134.1 | 4 - 5 | 1100 | 147 | |
| GenBank mRNA | AK058722.1 | - | 1 - 2 | 108 | 147 | |
| AK069367.1 | - | 4 - 5 | 1082 | 147 | ||
More Information
Set Membership: |
|

LOC4335881
Genomic Map