Assay Details
| Gene Symbol: LOC4336046 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778342.2 | XP_015633828.1 | 1 - 2 | 281 | 89 | |
| GenBank mRNA | AK102874.1 | - | 1 - 2 | 281 | 89 | |
| AK109192.1 | - | 1 - 2 | 281 | 89 | ||
More Information
Set Membership: |
|

LOC4336046
Genomic Map