Assay Details
| Gene Symbol: LOC4336046 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778342.2 | XP_015633828.1 | 4 - 5 | 524 | 123 | |
| XM_015778343.2 | XP_015633829.1 | 4 - 5 | 469 | 123 | ||
| GenBank mRNA | AK109192.1 | - | 4 - 5 | 524 | 123 | |
More Information
Set Membership: |
|

LOC4336046
Genomic Map