Assay Details
| Gene Symbol: LOC4336046 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778342.2 | XP_015633828.1 | 7 - 8 | 764 | 149 | |
| XM_015778343.2 | XP_015633829.1 | 7 - 8 | 709 | 149 | ||
| GenBank mRNA | AK102874.1 | - | 7 - 8 | 764 | 149 | |
| AK109192.1 | - | 7 - 8 | 764 | 149 | ||
More Information
Set Membership: |
|

LOC4336046
Genomic Map