Assay Details
| Gene Symbol: LOC4336249 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780578.2 | XP_015636064.1 | 4 - 5 | 1615 | 105 | |
| XM_015780580.2 | XP_015636066.1 | 3 - 4 | 1397 | 105 | ||
| GenBank mRNA | AK101116.1 | - | 4 - 5 | 1609 | 105 | |
More Information
Set Membership: |
|

LOC4336249
Genomic Map