Assay Details
| Gene Symbol: LOC4336275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781035.2 | XP_015636521.1 | 4 - 5 | 2437 | 131 | |
| XM_015781036.2 | XP_015636522.1 | 4 - 5 | 2469 | 131 | ||
| XM_015781037.2 | XP_015636523.1 | 4 - 5 | 2376 | 131 | ||
| GenBank mRNA | AK060753.1 | - | 3 - 4 | 1514 | 131 | |
| AK103154.1 | - | 4 - 5 | 2311 | 131 | ||
More Information
Set Membership: |
|

LOC4336275
Genomic Map