Assay Details
| Gene Symbol: LOC4336696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781679.2 | XP_015637165.1 | 2 - 3 | 681 | 75 | |
| GenBank mRNA | AK064998.1 | - | 2 - 3 | 689 | 75 | |
More Information
Set Membership: |
|

LOC4336696
Genomic Map